
Etonogestrel

Synonym(s):
(17α)-13-Ethyl-17-hydroxy-11-methylene-18,19-dinorpregn-4-en-20-yn-3-one, 3-Keto-desogestrel, 3-Oxo-desogestrel, Etonogestrel
Empirical Formula (Hill Notation):
C22H28O2
CAS Number:
Molecular Weight:
324.46
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3