Skip to Content
Merck
Ethyl acetoacetate-3,4-13C2 endotoxin tested, 99 atom % 13C

Ethyl acetoacetate-3,4-13C2

Ethyl acetoacetate-3,4-13C2 endotoxin tested, 99 atom % 13C
Linear Formula:
13CH313COCH2CO2CH2CH3
CAS Number:
Molecular Weight:
132.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing