Skip to Content
Merck
(±)-Methamphetamine-D8 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

(±)-Methamphetamine-D8 solution

(±)-Methamphetamine-D8 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C10D8H7N
CAS Number:
Molecular Weight:
157.28

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique