Skip to Content
Merck
Dimethenamid PESTANAL®, analytical standard

Dimethenamid

Dimethenamid PESTANAL®, analytical standard
Synonym(s):
2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-(2-methoxy-1-methylethyl)acetamide
Empirical Formula (Hill Notation):
C12H18ClNO2S
CAS Number:
Molecular Weight:
275.79

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique