
Cyclo(His-Pro)

Synonym(s):
CHP
Empirical Formula (Hill Notation):
C11H14N4O2
CAS Number:
Molecular Weight:
234.25
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing