
CX93

Synonym(s):
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Pd(dppf)Cl2
Linear Formula:
(C17H14P)2Fe · PdCl2
CAS Number:
Molecular Weight:
731.70
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing