
Chlorpheniramine Impurity A

Synonym(s):
2-(4-Chlorophenyl)-4-(dimethylamino)-2-[2-(dimethylamino)ethyl]butanenitrile
Empirical Formula (Hill Notation):
C16H24ClN3
CAS Number:
Molecular Weight:
293.83
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing