
Carmoisine

Synonym(s):
Chromotrope FB, Acid Red 14, Disodium 4-hydroxy-3-[(4-sulfo-1-naphthalenyl)azo]-1-naphthalenesulfonate, Mordant Blue 79
Empirical Formula (Hill Notation):
C20H12N2Na2O7S2
CAS Number:
Molecular Weight:
502.43
Products
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing