
Buspirone Related Compound N

Synonym(s):
8,8′-(Butane-1,4-diyl)bis(8-azaspiro[4.5]decane-7,9-dione)
Empirical Formula (Hill Notation):
C22H32N2O4
CAS Number:
Molecular Weight:
388.50
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing