
Bis(7-methyloctyl) phthalate

Synonym(s):
DINP, Diisononyl phthalate, Phthalic acid bis(7-methyloctyl) ester
Empirical Formula (Hill Notation):
C26H42O4
CAS Number:
Molecular Weight:
418.61
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing