
Azelastine Related Compound B

Synonym(s):
N′-(1-Methylazepan-4-yl)benzohydrazide hydrochloride, 1-Methyl-4-(2-benzoylhydrazino)azapan hydrochloride
Empirical Formula (Hill Notation):
C14H21N3O · HCl
CAS Number:
Molecular Weight:
283.80
Products
Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2