
Amlodipine Related Compound K

Synonym(s):
Ethyl 6-(2-chlorophenyl)-8-methyl-3,4,6,7-tetrahydro-2H-benzo[b][1,4]oxazine-5-carboxylate
Empirical Formula (Hill Notation):
C18H20ClNO3
CAS Number:
Molecular Weight:
333.81
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2