
Amifostine trihydrate

Synonym(s):
2-(3-Aminopropyl)aminoethyl phosphorothioate, WR2721
Empirical Formula (Hill Notation):
C5H15N2O3PS · 3 H2O
CAS Number:
Molecular Weight:
268.27
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing