Amido black 10 B (C.I.20470)
Synonym(s):
Amido black 10 B (C.I.20470)
Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing