
Allopurinol Related Compound A

Synonym(s):
3-Amino-4-pyrazolecarboxamide hemisulfate salt
Linear Formula:
(C4H6N4O)2 · H2SO4
CAS Number:
Molecular Weight:
350.31
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2