
Acetaldehyde-2,4-DNPH

Synonym(s):
Acetaldehyde-2,4-dinitrophenylhydrazone
Linear Formula:
CH3CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
224.17
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2