
6-Chloro-4-(2-chlorophenyl)quinazoline-2-carboxaldehyde

Synonym(s):
6-Chloro-4-(2-chlorophenyl)-2-quinazolinecarboxaldehyde
Empirical Formula (Hill Notation):
C15H8Cl2N2O
CAS Number:
Molecular Weight:
303.14
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing