
4-Fluoro-7-sulfamoylbenzofurazan

Synonym(s):
4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide, ABD-F
Empirical Formula (Hill Notation):
C6H4FN3O3S
CAS Number:
Molecular Weight:
217.18
Products
Product Category
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing