
4-Benzylpyridine

Empirical Formula (Hill Notation):
C12H11N
CAS Number:
Molecular Weight:
169.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing