
27-hydroxycholesterol

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing