Key Documents
Z756911
Universal animal restrainer
small, pack of 1 ea
Synonym(s):
animal container
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
material
clear acrylic
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H46400-0001
General description
Holds animal: mice
A variety of small animals and rodents can be restrained with this unit simply by moving the head gate to one of 4 different positions. To accommodate larger rodents, simply pull out the inner floor, increasing the available room. Stable base legs are provided by milled, chrome-plated brass thumb screws
  • Humanely restrain small animals
  • Comes with vented head gate and tail gate that are easily removed
  • Occlusion cuff held close to the animal′s body with the tailgate
Animal weight: 10-40 g
Minimum restraining: 5.1 x 2.9 cm (2 x 1 1/8 in.)
Maximum restraining: 10.2 x 2.9 cm (4 x 1 1/8 in.)