Key Documents
MTCP5212200U
MTC Bio ProPette LE 12-Channel Adjustable Pipettor
capacity 20-200 μL
Synonym(s):
pipette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.15
feature
adjustable
manual
no. of channels 12
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio P5212-200U
capacity
20-200 μL
General description
Ideal for all 96-well applications, Propette LE Multi-Channel Pipettes are lightweight, rugged, and capable of standing up to the demands of everyday use in a busy laboratory environment.
ProPette LE 12 -Channel Pipette, 20 to 200µL
Features and Benefits
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (1.0µL-300µL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)