Skip to Content
Merck
All Photos(1)

Key Documents

GPNT

9061601, rat brain, Fusiform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

GPNT, from rat brain, 09061601

biological source

rat brain

growth mode

Adherent

karyotype

Not specified

morphology

Fusiform

products

GPNT cells express tight junction proteins

receptors

P-glycoprotein, PECAM-1, ICAM-1, VCAM-1 Glut-1, transferrin receptor, MHC class I

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

Cell Line Origin

An immortalised Lewis rat brain vascular endothelial cell line

Cell Line Description

GPNT was derived from the immortalised rat brain endothelial cell line GP8 by lipofectin-mediated transfection with a vector containing the puromycin resistance gene. GPNT exhibits a non-transformed phenotype.

DNA Profile

Not specified

Culture Medium

Hams F10 + 10% Heat-inactivated FCS + 2 mM Glutamine + 2 ng/ml basic fibroblast growth factor + 80 μg/ml heparin. All plasticware is pre-coated for several hours with collagen (Type I from calf skin) at 10μg/cm2 from a 0.01% solution in acetic acid. After removal the coated plasticware is exposed to a mMonia vapour for 5 min. Plasticware must be rinsed thoroughly with PBS prior to use.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 , i.e., seeding at 3 x 10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Population doubling approx 10hrs.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service