Skip to Content
Merck
All Photos(1)

Key Documents

PHR2308

Loratadine Related Compound B

Synonym(s):

8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21ClN2
CAS Number:
Molecular Weight:
324.85
UNSPSC Code:
41116107
NACRES:
NA.24

Agency

traceable to USP 1370291

Quality Level

API family

loratadine

form

solid

application(s)

pharmaceutical small molecule

storage temp.

2-8°C

InChI

1S/C20H21ClN2/c1-23-11-8-14(9-12-23)19-18-7-6-17(21)13-16(18)5-4-15-3-2-10-22-20(15)19/h2-3,6-7,10,13H,4-5,8-9,11-12H2,1H3

InChI key

VLXSCTINYKDTKR-UHFFFAOYSA-N

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service