Skip to Content
Merck
All Photos(2)

Key Documents

MABF112

Sigma-Aldrich

Anti-CD70 Antibody, clone TAN 1-7

clone TAN 1-7, from rat

Synonym(s):

CD70 antigen, CD27 ligand, CD27-L, Tumor necrosis factor ligand superfamily member 7, CD70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

TAN 1-7, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd70(21948)

General description

CD70 is an interesting membrane bound cellular factor that acts as a cytokine which binds the receptor CD27. Binding induces the proliferation and co-stimulation of T cells and enhances the generation of cytolytic T-cells. Thus, CD70 plays important roles in T cell adaptive T cell response as well as in innate immunity and is required for the proper activation of the anti-viral adaptive response. CD70 also serves as a regulatory role in early cytokine responses to pathogens through the maintenance of Treg cell survival and function. Recent research also indicates that CD70 expression could be important in autoimmune disease regulation and in treatment of autoimmune diseases such as rheumatoid arthritis. CD70 is expressed on T cells, splenocytes, and thymocytes and it belongs to the TNF family of cellular signaling proteins.

Immunogen

Epitope: Extracellular domain
Recombinant protein corresponding to the extracellular domain of mouse CD70.

Application

Anti-CD70 Antibody, clone TAN 1-7 | MABF112 is an antibody against CD70 Antibody for use in Western Blotting, Flow Cytometry.
Flow Cytometry Analysis: 2 µg of this antibody detected CD70 in mouse bone marrow dendritic cells treated with 2000U/mL GM-CSF and 0.1 ug/mL of LPS.
Functional Blocking Analysis: A representative lot detected CD70 in splenic CD8a+ DCs (Arora, P., et al. (2014). Immunity. 40:105-116).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in mouse spleen tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected CD70 in 10 µg of mouse spleen tissue lysate.

Target description

~27 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service