MAB858-1P-KC
Anti-Respiratory Syncytial Virus, Fusion, clone 133-1H (1mg) KC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
Recommended Products
antibody form
purified antibody
Quality Level
clone
133-1H
monoclonal
species reactivity
human, mouse
technique(s)
ELISA: suitable
immunofluorescence: suitable
Storage and Stability
Refrigerator (2–8°C)
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service