Skip to Content
Merck
All Photos(1)

Key Documents

MAB8200

Sigma-Aldrich

Anti-HSV-2 Antibody, clone 3C5.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody product type

primary antibodies

species reactivity

virus

packaging

vial of 100 μg

manufacturer/tradename

Millipore

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

shipped in

2-8°C

storage temp.

2-8°C

Application

Tested by Immunohistochemistry.

Features and Benefits

Monoclonal antibody Anti-Herpes Simplex Virus 2, clone 3C5.2 (Cat. No. MAB8200) is tested for use in Immunohistochemistry, Immunofluorescence and Western Blot for the detection of HSV-2.

Target description

Herpes simplex virus type 2 (HSV-2) is almost exclusively sexually transmitted, causing infection in the genital or anal area (genital herpes).

Physical form

Purified immunoglobulin in 0.02M phosphate buffer, pH 7.6, 0.25M sodium chloride containing 0.1 sodium azide.

Storage and Stability

Store at +2°C to +8°C. It is recommended to store the product undiluted and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service