Skip to Content
Merck
All Photos(1)

Key Documents

1.93271

Supelco

Potassium hydrogen phthalate

for analysis EMPARTA® ACS

Synonym(s):

Potassium hydrogen phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5KO4
CAS Number:
Molecular Weight:
204.22
MDL number:

grade

ACS reagent

storage temp.

room temp

InChI

1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1

InChI key

IWZKICVEHNUQTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93271.0521: 500 g, Minimum order quantity: 20

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

387.1 °F - Pensky-Martens closed cup

Flash Point(C)

197.3 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service