Merck
HomeMammalian Cell CultureNutrient Mixture F-12 Ham Formulation
Nutrient Mixture F-12 Ham Formulation
Ham's nutrient mixtures were originally developed to cultivate Chinese hamster ovary (CHO) cells, mouse L-cells, and HeLa cells. These mixtures were formulated for use with or without serum supplementation like fetal bovine serum depending on the cell type cultured, and does not contain growth factors.
Ham's F-12 Nutrient Mixture, a modification on Ham’s F10, has been used to grow CHO cells, primary rat hepatocytes, and rat prostate epithelial cells. Ham's F-12 has several important modifications - Coon's modification, developed for culturing hybrid cells produced by viral fusion, and Kaighn's modification, which has increased concentrations of select amino acids, pyruvate, and modified salts. Kaighn's modification was designed to support the growth of differentiated rat and chicken cells and primary human hepatocytes. Coon’s modification consists of 2x the standard concentration of amino acids and pyruvate and includes ascorbic acid and adjusted salt concentrations. The Coon’s formula also contains 0.863 mg/L zinc sulfate, which may render it unsuitable for culturing mouse L-cells.
Nutrient Mixture F-12 Ham
ComponentN4888
[1x] g/L
N6658
[1x] g/L
N6760
g/L
Inorganic Salts
Calcium Chloride0.03330.03330.0333
Cupric Sulfate • 5H2O0.00000250.00000250.0000025
Ferrous Sulfate • 7H2O0.0008340.0008340.000834
Magnesium Chloride0.05760.05760.0576
Potassium Chloride0.2240.2240.224
Sodium Bicarbonate1.1761.176
Sodium Chloride7.5997.5997.599
Sodium Phosphate Dibasic (anhydrous)0.142040.142040.14204
Zinc Sulfate • 7H2O0.0008630.0008630.000863
Amino Acids
L-Alanine0.0090.0090.009
L-Arginine • HCl0.2110.2110.211
L-Asparagine • H2O0.015010.015010.01501
L-Aspartic Acid0.01330.01330.0133
L-Cysteine • HCl • H2O0.0350.0350.035
L-Glutamic Acid0.01470.01470.0147
L-Glutamine0.1460.146
Glycine0.007510.007510.00751
L-Histidine • 3HCl • H2O0.020960.020960.02096
L-Isoleucine0.003940.003940.00394
L-Leucine0.01310.01310.0131
L-Lysine • HCl0.03650.03650.0365
L-Methionine0.004480.004480.00448
L-Phenylalanine0.004960.004960.00496
L-Proline0.03450.03450.345
L-Serine0.01050.01050.0105
L-Threonine0.01190.01190.0119
L-Tryptophan0.002040.002040.00204
L-Tyrosine • 2Na • 2H2O0.007780.007780.00778
L-Valine0.01170.01170.0117
Vitamins
D-Biotin0.00000730.00000730.000073
Choline Chloride0.013960.013960.01396
Folic Acid0.001320.001320.00132
myo-Inositol0.0180.0180.018
Niacinamide0.0000370.0000370.000037
D-Pantothenic Acid (hemicalcium)0.000480.000480.00048
Pyridoxine • HCl0.0000620.0000620.000062
Riboflavin0.0000380.0000380.000038
Thiamine • HCl0.000340.000340.00034
Vitamin B120.001360.001360.00136
Other
D-Glucose1.8021.8021.802
Hypoxanthine0.004080.004080.00408
Linoleic Acid0.0000840.0000840.000084
Phenol Red • Na0.00130.00130.0013
Putrescine • HCl0.0001610.0001610.000161
Pyruvic Acid • Na0.110.110.11
Thioctic Acid0.000210.000210.00021
Thymidine0.000730.000730.00073
Add
L-Glutamine0.146
Sodium Bicarbonate1.176
Nutrient Mixture F-12 Ham HEPES Modification
ComponentN8641
[1x] g/L
Inorganic Salts
Calcium Chloride0.0333
Cupric Sulfate • 5H2O0.0000025
Ferrous Sulfate • 7H2O0.000834
Magnesium Chloride0.0576
Potassium Chloride0.224
Sodium Bicarbonate1.176
Sodium Chloride7.1
Sodium Phosphate Dibasic (anhydrous)0.14204
Zinc Sulfate • 7H2O0.000863
Amino Acids
L-Alanine0.009
L-Arginine • HCl0.211
L-Asparagine • H2O0.01501
L-Aspartic Acid0.0133
L-Cysteine • HCl • H2O0.035
L-Glutamic Acid0.0147
Glycine0.00751
L-Histidine • 3HCl • H2O0.02096
L-Isoleucine0.00394
L-Leucine0.0131
L-Lysine • HCl0.0365
L-Methionine0.00448
L-Phenylalanine0.00496
L-Proline0.0345
L-Serine0.0105
L-Threonine0.0119
L-Tryptophan0.00204
L-Tyrosine • 2Na • 2H2O0.00778
L-Valine0.0117
Vitamins
D-Biotin0.0000073
Choline Chloride0.01396
Folic Acid0.00132
myo-Inositol0.018
Niacinamide0.000037
D-Pantothenic Acid (hemicalcium)0.000238
Pyridoxine • HCl0.000062
Riboflavin0.000038
Thiamine • HCl0.00034
Vitamin B120.00136
Other
D-Glucose1.802
HEPES5.958
Hypoxanthine0.00408
Linoleic Acid0.000084
Phenol Red • Na0.0013
Putrescine • HCl1.000161
Pyruvic Acid • Na0.11
Thioctic Acid0.00021
Thymidine0.00073
Add
L-Glutamine0.146
Nutrient Mixture F-12 Ham Kaighn's Modification
ComponentN3520
[1x] g/L
Inorganic Salts
Calcium Chloride0.10207
Cupric Sulfate • 5H2O0.0000025
Ferrous Sulfate • 7H2O0.000834
Magnesium Chloride0.04961
Magnesium Sulfate0.19264
Potassium Chloride0.28329
Potassium Phosphate Monobasic0.058523
Sodium Phosphate Dibasic0.11502
Sodium Chloride7.5972
Zinc Sulfate • 7H2O0.00014375
Amino Acids
L-Alanine0.017818
L-Arginine0.4214
L-Asparagine • H2O0.03002
L-Aspartic Acid0.02662
L-Cysteine • HCl • H2O0.07024
L-Glutamic Acid0.02942
L-Glutamine0.2922
Glycine0.015014
L-Histidine • HCl • H2O0.04192
L-Isoleucine0.007872
L-Leucine0.02624
L-Lysine • HCl0.07304
L-Methionine0.008952
L-Phenylalanine0.009912
L-Proline0.06906
L-Serine0.02102
L-Threonine0.02382
L-Tryptophan0.004084
L-Tyrosine0.010872
L-Valine0.02342
Vitamins
D-Biotin0.00007329
Choline Chloride0.01396
Folic Acid0.0013242
Hypoxanthine0.004083
myo-Inositol0.01802
Niacinamide0.00003663
D-Pantothenic Acid (hemicalcium)0.000477
Putrescine • 2HCl0.0003222
Pyridoxine • HCl0.00006168
Riboflavin0.00003764
Thiamine • HCl0.0003373
Thymidine0.0007266
Vitamin B120.0013554
Other
D+-Glucose1.26
Phenol Red • Na0.00331806
Pyruvic Acid • Na0.22
DL-6,8-Thioctic Acid0.0002063
Add
Sodium Bicarbonate2.5
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch