DiscoveryBIO Wide Pore C18
Discovery BIO Wide Pore C18
HPLC Columns and Capillaries
Bonded Phase: Covalently-bonded octadecylsilane (C18), endcapped
Silica: Spherical, 3µM, 5µM and 10µM diameters, 300Å pore diameter, high purity (<10ppm metals)
Solutions for Protein and Peptide Separation Challenges
- Proteomics and biotherapeutics: high resolution of synthetic peptides and peptide maps
- LC/MS; no-bleed, capillary dimensions, and pH stability
- Peptide purification: Scalable from analytical to preparative
- Reproducible separations batch-to-batch and column-to-column
Here's the Proof
Matched Selectivity from Analytical to Prep
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 3µM; 15cm x 4.6mm, 5µM; 15cm x 10mm, 10µM, Mobile Phase: (A) 80:20, (0.1% TFA in H2O) : (0.1%TFA in MeCN), (B) 66:34, (0.1% TFA in H2O) : (0.1%TFA in MeCN), Flow Rate: 6.02cm/min, Temp: 30 °C, Detection: 215nm, Sample: Peptide Mix (Cat. No. P2693)
Column Parameters & Run Conditions: |
---|
![su-g001511-13 su-g001511-13](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-g001511-13.gif)
Gradient: |
---|
Peptide Map of Carboxymethylated Apohemoglobin Tryptic Digest
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 5µm; Mobile Phase: (A) 95:5, (0.1% TFA in H2O):(0.1% TFA in MeCN); (B) 50:50, (0.1% TFA in H2O):(0.1% TFA in MeCN); Flow Rate: 1mL/min; Temp: 30 °C; Detection: 215nm; Injection: 50µL; Sample: caboxymethylated apohemoglobin tryptic digest; Gradient: 0-100% B in 65 min
![su-g001510](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-g001510.gif)
Stability of Discovery Bio Wide Pore C18 at Alkaline pH
Alkaline pH Stability
Retention times of the sample components remained constant after 29,000 bed volumes were passed through the column.
Column: Discovery BIO Wide Pore C18, 5cm x 4.6mm, 5µM; Mobile Phase: 25mM potassium phosphate, pH 8 : MeOH (95:5); Flow Rate: 2mL/min; Temp: 35 °C; Detection: 254nm
![su-g001520 su-g001520](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-g001520.gif)
Undetectable LC/MS Bleed
LC/MS Bleed Analysis on Discovery BIO Wide Pore C18
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 3µM; Mobile Phase: (A) 0.1% TFA in H2O, (B) 0.1% TFA in MeOH; Flow Rate: 1mL/min; Temp: 30 °C; Gradient: 0-100% B in 15 min, hold for 5 min, then back to 0% B for 10 min
No TIC baseline rise relative to blank run (gradient run w/o column)
![su-g001522 su-g001522](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-g001522.gif)
![su-g001523](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-g001523.gif)
Lot to Lot Reproducibility of Discovery Bio Wide Pore c18
![su-g001514-6 su-g001514-6](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-g001514-6.gif)
Column: Discovery BIO Wide Pore C18, 15cm x 4.6mm, 5µM; Mobile Phase: 80% 10mM ammonium acetate, pH 6.8 : 20% MeOH; Flow Rate: 1mL/min; Temp: 35 °C; Detection: 254nm; Injection: 5µL
- Uracil (8µg/mL)
- Procainamide (30µg/mL)
- Sorbic acid (4µg/mL)
- Pyridine (100µg/mL)
- Caffeine (40µg/mL)
- Phenol (240µg/mL)
Ordering Information
![su-discovery-bio su-discovery-bio](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/su-discovery-bio.gif)