Merck
UHPLC Gradient Analysis of Boswellic Acids on Titan C18
UHPLC Gradient Analysis of Boswellic Acids on Titan C18 application for UHPLC
Materials
mobile phase component
Product No.
Description
Pricing
Phosphoric acid
≥85 wt. % in H2O, ≥99.999% trace metals basis
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 0.1% phosphoric acid; [B] acetonitrile
gradient
40% B for 0.5 min; to 100% B in 2 min; held at 100% B for 10 min
flow rate
0.4 mL/min
pressure
4050 psi (279 bar)
column temp.
35 °C
detector
UV, 210 nm
injection
2 μL
sample
6 Boswellic Acids 50μg/mL in 70:30, water:methanol
Description
Legal Information
null