UHPLC Analysis of Dimethyl Sulfoxide and Dimethylformamide on Ascentis® Express HILIC
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.0 μm particles (51406-U)
mobile phase
[A] 0.1% ammonium hydroxide; [B] acetonitrile; (5:95, A:B)
flow rate
0.4 mL/min
pressure
(205 bar)
column temp.
35 °C
detector
MS, ESI(+), MRM, 73.95/45.97, 78.85/63.92
injection
0.5 μL
sample
0.5 μg/mL each in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany