Merck
TLC Analysis Of A Natural Product From Cosmetical Matrix on HPTLC Silica gel 60 NH2 F254s
TLC Analysis Of A Natural Product From Cosmetical Matrix on HPTLC Silica gel 60 NH2 F254s application for TLC
Materials
accessory
Product No.
Description
Pricing
Bottle-type sprayer
capacity 240 mL
Rectangular TLC developing tanks, complete
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Sigma TLC spray box
Spectroline® E-Series UV lamp
output8 W, AC/DC input 115 V AC
Rectangular TLC developing tanks, complete
L × H × W 7.5 cm × 15.5 cm × 8.0 cm
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),
coated tip
Spectroline® E-Series UV lamp
output6 W, AC/DC input 120 V AC
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),
coated tip
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
Spectroline® E-Series UV lamp
output6 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output4 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output8 W, AC/DC input 230 V AC
Latch-lid TLC developing chambers
For use with 10 cm × 10 cm plates
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Latch-lid TLC developing chambers
For use with 20 cm × 20 cm plates
Spectroline® E-Series UV lamp
output4 W, AC/DC input 115 V AC
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
capacity 20 μL, accuracy: ±0.5%
Reagent sprayer for TLC plates
volume 250 mL, Bottle-Type
CONDITIONS
sample preparation
Standard was diluted in 10 mL isopropanol, sample was directly applied to the plate by CAMAG ATS4 6 mm bandwise.
plate
1.15161 HPTLC Silica gel 60 RP-18 F 254s MS-grade 20cm x 10 cm (1.151161.0001)
eluent
[A] methanol; [B] acetonitrile; (9:1, A:B)
migration distance
6 cm
chamber
normal chamber without chamber saturation
sample
0.150 μL
detector
UV, 254 nm (Identification by coupling the TLC – MS Interface (CAMAG) with the Advion expression) (CMS mass spectrometer (+)-ESI mode. Eluent: Acetonitrile / Water 95:5 + 0.1% TFA.)