Merck
US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18
US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® Cyano HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
Discovery® Cyano HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Ascentis® C18 HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
Ascentis® C18 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Ascentis® C8 HPLC Column
3 μm particle size, L × I.D. 15 cm × 4.6 mm
Ascentis® C8 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
Ascentis® Express C8, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
Coupler for Legacy Guard Column Holder
PEEK, I.D. × O.D. × Overall L 0.010 in. × 1/16 in. × 1 in.
Supelguard Guard Cartridge Holder
Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)
guard cartridge
Product No.
Description
Pricing
Discovery® Cyano Supelguard Cartridge
5 μm particle size, L × I.D. 2 cm × 4 mm
Ascentis® C18 Supelguard Guard Cartridge
5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Ascentis® C18 Supelguard Guard Cartridge
3 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Ascentis® C8 Supelguard Guard Cartridge
5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
CONDITIONS
column
Ascentis C18, 25 cm x 2.1 mm I.D., 5 μm particles (581305-U)
mobile phase
(A) water: (B) methanol; (50:50, A:B)
flow rate
0.2 mL/min
column temp.
60 °C
detector
UV, 254 nm
injection
1 μL
sample
acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany