HPLC Analysis of Warfarin Enantiomers on Astec® CYCLOBOND I 2000 (UV at 254 nm)
![HPLC Analysis of Warfarin Enantiomers on Astec® CYCLOBOND I 2000 (UV at 254 nm) application for HPLC](/deepweb/assets/sigmaaldrich/product/images/149/326/fff19d85-9cc1-4b85-9dd0-2923395236a6/800/fff19d85-9cc1-4b85-9dd0-2923395236a6.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CYCLOBOND I 2000, 25 cm × 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] acetonitrile: [B] acetic acid: [C] triethylamine; (100:0.3:0.25, A:B:C)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 254 nm
injection
5 μL
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null
null