Merck
HPLC Analysis of N-Linked Glycans on BIOshell Glycan
HPLC Analysis of N-Linked Glycans on BIOshell Glycan application for HPLC
Materials
analytical column
Product No.
Description
Pricing
BIOshell Glycan HPLC Columns
L × I.D. 10 cm × 2.1 mm, 2.7 μm particle size
CONDITIONS
column
BIOshell Glycan HPLC
Column (50993-U)
mobile phase
(A) Water with 100 mM ammonium formate, pH 4.5; (B) Acetonitrile
gradient
min, %B, Flow (mL/min): 0-74-0.4; 15-71-0.4; 25-55-0.4; 26-45-0.2; 30-45-0.2; 31-75-0.4; 35-75-0.4.
pressure
1130 psi (78 bar)
column temp.
50 °C
detector
Fluorescence (&#955ex = 260 nm, &#955em = 430 nm)
injection
3 μL
sample
in 70% mobile phase B
Description
Analysis Note
Separation pf a mix of 2-AB labled N-linked glycans released from human IgG was optimized on BIOshell Glycan HPLC Columns. Baseline resolutions for all of the individual glycans were observed in both mixes, except for the positional isomer pair G1F and G1F′ which were partially resolved. 
Legal Information
null