HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP, POM Conditions
![HPLC Analysis of Mianserin Enantiomers on Astec® Cellulose DMP, POM Conditions application for HPLC](/deepweb/assets/sigmaaldrich/product/images/643/469/59f88b8d-6d48-452f-89f9-7ad05aec7b75/800/59f88b8d-6d48-452f-89f9-7ad05aec7b75.jpg)
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] methanol; [B] DEA; (100:0.1, A:B)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
inj. temp.
2 μL
sample
2 mg/mL in mobile phase