HPLC Analysis of β-Lactam Antibiotics on Discovery® RP-Amide C16
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery RP-Amide C16, 15 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
[A] 0.01% TFA in water; [B] 0.01% TFA in acetonitrile
flow rate
1.5 mL/min
column temp.
30 °C
detector
UV, 254 nm
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany