Merck
HPLC Analysis of Fluoroquinolone Antibiotics on Ascentis® Phenyl
HPLC Analysis of Fluoroquinolone Antibiotics on Ascentis® Phenyl application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Phenyl HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 μm particles (581616-U)
mobile phase
[A] 13 mM ammonium formate (pH 3.0 with concentrated formic acid): [B] acetonitrile; (80:20, A:B)
flow rate
1 mL/min, split to the MS
column temp.
35 °C
detector
MS, ESI (+) in selected ion recording (SIR) mode
injection
5 μL
sample
as indicated in 80:20, 13 mM ammonium formate (pH 3.0 with concentrated formic acid):acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany