Merck
HPLC Analysis of AQC-Alanine Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of AQC-Alanine Enantiomers on Astec® CHIROBIOTIC® T2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
DL-Alanine
≥99% (HPLC)
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (50:50, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
0.5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC