Merck
HPLC Analysis of Amino Acids on Ascentis® Si (Silica), HILIC Mode
HPLC Analysis of Amino Acids on Ascentis® Si (Silica), HILIC Mode application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Si HPLC Column
5 μm particle size, L × I.D. 15 cm × 2.1 mm
CONDITIONS
column
Ascentis Si, 15 cm × 2.1 mm I.D., 5 μm particles (581509-U)
mobile phase
[A] 100 mM ammonium formate (pH 3.0, with formic acid), [B] water, [C] acetonitrile
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI (+), full scan
injection
2 μL
sample
in 10:90, (50 mM ammonium formate/formic acid, pH 3.0):acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany