Merck
HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® T (No Mobile Phase Modifier)
HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® T (No Mobile Phase Modifier) application for HPLC
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] acetonitrile; (50:50, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 205 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC