Merck
US EPA Method 524.2: GC Analysis of Volatiles on SPB-624 after Purge & Trap using "K" Trap
US EPA Method 524.2: GC Analysis of Volatiles on SPB-624 after Purge & Trap using "K" Trap suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-624 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.40 μm
related product
Product No.
Description
Pricing
Purge/Trap K Vocarb® 3000
for use with OI Analytical Eclipse 4660, 4560
standard
Product No.
Description
Pricing
EPA 524.2 Rev 4 Update Mix
2000 μg/mL in methanol, analytical standard
EPA 524 Internal Standard Mix
certified reference material, 2000 μg/mL each component in methanol
EPA 524.2 Rev 4 Update Ketones Mix
certified reference material, 2000 μg/mL in methanol: water (9:1)
EPA 502/524 Volatiles Organic Calibration Mix A (without gases)
certified reference material, 200 μg/mL each component in methanol, ampule of 1 mL
Volatile Organic Compounds Mix 6
certified reference material, 2000 μg/mL each component in methanol
CONDITIONS
sample/matrix
each analyte at 10 ppb in 25 mL water
purge trap
VOCARB 3000 "K" (24940-U)
dry purge
2 min
desorption preheat
205 °C
desorption process
40 mL/min at 210 °C for 2 min
bake
260 °C for 10 min
transfer line/valve temp.
110 °C
sample preparation
purge is 40 mL/min at 25 °C for 11 min
column
SPB-624, 30 m x 0.25 mm I.D., 1.4 μm (24255)
oven
40 °C (2 min), 7 °C/min to 135 °C, 30 °C/min to 230 °C (3 min)
carrier gas
helium, 1.2 mL/min
injection
30:1 split
liner
0.75 mm I.D. SPME
Description
Analysis Note
Method 524.2 describes the analysis of volatile organic compounds in surface, ground, and drinking water. This application illustrates the analysis of the compounds listed in this method on a 30m x 0.25 mm I.D., 1.4 μm SPB-624 column using purge and trap / GC-MS.