GC Analysis of Fuel Oil #4 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
![GC Analysis of Fuel Oil #4 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC](/deepweb/assets/sigmaaldrich/product/images/156/377/f336dc6f-553b-4641-baea-599cbc7c2144/800/f336dc6f-553b-4641-baea-599cbc7c2144.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
carrier gas
hydrogen, 45 cm/sec constant
column
Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
detector
FID, 350 °C
inj. temp.
250 °C
injection
0.3 μL, 200:1 split, 0.02 min pre-injection dwell time
liner
2 mm I.D., straight
oven
80 °C, 50 °C/min to 325 °C
sample
No.4 Fuel Oil standard, 50 mg/mL in hexane (47535-U)
Description
Legal Information
null