US EPA Method 608/8081/OLM04.2 PEST: GC Analysis of Organochlorine Pesticides on Equity-1701 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
CONDITIONS
column
Equity-1701, 15 m × 0.10 mm I.D., 0.10 μm (28343-U)
oven
100 °C, 25 °C/min to 280 °C
inj. temp.
225 °C
detector
ECD, 300 °C
carrier gas
hydrogen, 40 cm/sec constant
injection
2 μL, splitless (0.75 min)
liner
4 mm I.D., single taper
sample
50 ppb of a 22 component chlorinated pesticide standard in n-hexane
Description
Legal Information
null