Merck
GC Analysis of Solvents on a Packed Column (3% SP-1500 on Carbopack B)
GC Analysis of Solvents on a Packed Column (3% SP-1500 on Carbopack™ B) suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Coated GC Packing
phase SP®-1000, 0.1%, matrix 80/100 Carbopack C support, bottle of 15 g
CONDITIONS
column
10 ft (3.048m) × 1/8 in.(9.52 mm) stainless steel
packing
Carbopack B/3% SP®-1500
inj. temp.
0.3 μL 3% each component by volume (approx. 12 μg each)
column temp.
A & C - 70 °C to 225 °C at 4 °C/min B - 100 °C to 225 °C at 8 °C/min
detector
FID
carrier gas
nitrogen, 20 mL/min
Description
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
SP is a registered trademark of Sigma-Aldrich Co. LLC