Merck
GC Analysis of Formalin Solution on Carboxen®-1006 PLOT
GC Analysis of Formalin Solution on Carboxen®-1006 PLOT suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Nukol Capillary GC Column
size × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
Carboxen-1006 PLOT
oven
200 °C
detector
TCD
carrier gas
3.0 mL/min
injection
0.5 μL
sample
Formalin solution
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany