GC Analysis of Fermentation Compounds on a Packed Column (6.6% Carbowax® 20M on 80/120 Carbopack™ B AW)
CONDITIONS
column
2 m × 2 mm I.D. glass
packing
6.6% Carbowax 20M on 80/120 Carbopack B AW
oven
80 °C, 4 °C/min to 200 °C
detector
FID
carrier gas
nitrogen
injection
1 μL
sample
40-60 ppm each analyte in water:ethanol (50:50)
Description
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
Carbowax is a registered trademark of Union Carbide