Merck
  • Effect of polymorphisms of the MTHFR and APOE genes on susceptibility to diabetes and severity of diabetic retinopathy in Brazilian patients.
Effect of polymorphisms of the MTHFR and APOE genes on susceptibility to diabetes and severity of diabetic retinopathy in Brazilian patients.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas (2006-07-25)
F I V Errera, M E R Silva, E Yeh, C M C Maranduba, B Folco, W Takahashi, A C Pereira, J E Krieger, M R Passos-Bueno
ABSTRACT
Diabetes mellitus (DM) is a highly prevalent complex genetic disorder. There has been a worldwide effort in the identification of susceptibility genes for DM and its complications, and the 5-10-methylenetetrahydrofolate reductase (MTHFR) and apolipoprotein-E (APOE) genes have been considered good candidate susceptibility genes to this condition. The objectives of the present study were to determine if the 677T MTHFR and epsilon2/epsilon3/epsilon4 APOE alleles are risk factors for DM and for severity of diabetic retinopathy (DR). A total of 248 individuals were studied: 107 healthy individuals and 141 diabetic patients (46 with type 1 diabetes and 95 with type 2 diabetes), who also had DR (81 with non-proliferative DR and 60 with proliferative DR). The polymorphisms were analyzed by PCR followed by digestion with restriction enzyme or the single-nucleotide primer extension method. No evidence of association between the 677TT genotype of MTHFR gene and DM [cases: TT = 10/95 (10.6%); controls: TT = 14/107 (13%)] or with severity of DR was observed [cases: TT = 5/60 (8.5%); controls: TT = 9/81 (11.1%); P > 0.05]. We also did not find evidence of an association between APOE alleles and proliferative DR (epsilon2, epsilon3 and epsilon4 in cases: 9, 76, and 15%, and in controls: 5, 88, and 12%, respectively) but the carriers of epsilon2 allele were more frequent among patients with type 2 DM and DR than in controls [cases: 15/95 (15.8%); controls: 7/107 (6.5%); P < 0.05]. Therefore, our results suggest that the epsilon2 allele/APOE might be a risk factor for diabetes in the Brazilian population.