Merck
  • Multiresidue method using SPME for the determination of various pesticides with different volatility in confined atmospheres.
Multiresidue method using SPME for the determination of various pesticides with different volatility in confined atmospheres.
Analytical and bioanalytical chemistry (2004-04-16)
Federico Ferrari, Astrid Sanusi, Maurice Millet, Michel Montury
ABSTRACT
An analytical method is described for assessing the vapour concentration of 11 pesticides (bioallethrin, chlorpyriphos methyl, folpet, malathion, procymidone, quintozene, chlorothalonil, fonofos, penconazole and trimethacarb) in confined atmospheres (e.g. a greenhouse after pesticide application). This study is a successful extension of a method previously developed by the authors for dichlorvos to much less volatile pesticides. Sampling was performed by using polydimethylsiloxane-solid phase micro-extraction (PDMS-SPME) fibres immersed in a 250-mL sampling flask through which air samples were dynamically pumped from the analysed atmosphere. After a 40-min sampling duration, samples were analysed by GC/MS. Calibration was performed from a vapour-saturated air sample. The linearity of the observed signal versus pesticide concentration in the vapour phase was proved from spiked liquid samples whose headspace concentrations were measured by using the proposed method. This procedure gave calibration curves with regression coefficients ( R(2)) greater than 0.98, and the repeatability of these measurements was found with RSDs of 1.9-7.6%. As a field application test, this analysis procedure was used for the determination of gaseous procymidone concentrations as a function of time in the atmosphere of an experimental 8-m(2) and 20-m(3) greenhouse. The pesticide was sprayed according to real cultivation conditions, and measurements were made for 80 h after application (8 measurements). The observed concentrations found ranged from 200 to 500 micro g m(-3), thus indicating the level of contamination of the air breathed by people in such working conditions.
Product Description
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
IonSense® PDMS/DVB SPE-it Tips, pkg of 96 ea
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler